missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Oxybis(benzenesulfonyl hydrazide), 95%, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4,4′-Oxybis(benzenesulfonyl hydrazide) is an organic hydrazide used as a blowing agent for sponge rubber and expanded plastics. Used as an additive to polymers to make sponge rubber cosmetic puffs, electrostatographic toners, buoyant seats, insulation and packing materials, adhesives, sealants, and leather substitutes.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
4,4′-Oxybis(benzenesulfonyl hydrazide) is an organic hydrazide used as a blowing agent for sponge rubber and expanded plastics. Used as an additive to polymers to make sponge rubber cosmetic puffs, electrostatographic toners, buoyant seats, insulation and packing materials, adhesives, sealants, and leather substitutes.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications
Color | White |
Quantity | 100 g |
UN Number | UN3226 |
Formula Weight | 358.39 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 4,4'-Oxybis(benzenesulfonyl hydrazide) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'