missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrophenylazo)resorcinol, 90+%

Product Code. 11434923
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434923 500 g 500g
11414923 25 g 25g
11424923 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434923 Supplier Thermo Scientific Alfa Aesar Supplier No. A12232.36
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 74-39-5 | C12H9N3O4 | 259.22 g/mol

It is a derivitized resorcinol dye compound with an absorption maximum of 432 nm. Used as a pH indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a derivitized resorcinol dye compound with an absorption maximum of 432 nm. Used as a pH indicator.

Solubility
Soluble in dilute sodium hydroxide. Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-39-5
Molecular Formula C12H9N3O4
Molecular Weight (g/mol) 259.22
MDL Number MFCD00007310
InChI Key BIFFKKLAAIGZBV-UHFFFAOYSA-N
Synonym p-nitrophenylazoresorcinol, p-diazoviolet, p-nitrobenzeneazoresorcinol, magneson i, magnezon i, 4-p-nitrophenylazo resorcinol, p-diazviolet, 2,4-dihydroxy-4'-nitroazobenzene, magneson, 4-4-nitrophenylazo resorcinol
PubChem CID 5717413
IUPAC Name (4E)-3-hydroxy-4-[(4-nitrophenyl)hydrazinylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point ∼185°C (decomposition)
Quantity 500 g
Beilstein 674709
Merck Index 14,5695
Solubility Information Soluble in dilute sodium hydroxide. Insoluble in water.
Formula Weight 259.22
Percent Purity 95%
Chemical Name or Material 4- (4-Nitrophenylazo)resorcinol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.