missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrophenyl)morpholine, 98%, Thermo Scientific™

Catalog Number 11471437
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
10389-51-2
C10H12N2O3
208.217
MFCD00023317
IAJDSUYFELYZCS-UHFFFAOYSA-N
4-4-nitrophenyl morpholine, morpholine, 4-4-nitrophenyl, p-morpholinonitrobenzene, 4-p-nitrophenyl morpholine, 4-morpholinyl nitrobenzene, 4-morpholinonitrobenzene, n-p-nitrophenyl-morpholine, morpholine, 4-p-nitrophenyl, n-4-nitrophenyl morpholine
25222
4-(4-nitrophenyl)morpholine
C1COCCN1C2=CC=C(C=C2)[N+](=O)[O-]
This item is not returnable. View return policy
10389-51-2
C10H12N2O3
208.217
MFCD00023317
IAJDSUYFELYZCS-UHFFFAOYSA-N
4-4-nitrophenyl morpholine, morpholine, 4-4-nitrophenyl, p-morpholinonitrobenzene, 4-p-nitrophenyl morpholine, 4-morpholinyl nitrobenzene, 4-morpholinonitrobenzene, n-p-nitrophenyl-morpholine, morpholine, 4-p-nitrophenyl, n-4-nitrophenyl morpholine
25222
4-(4-nitrophenyl)morpholine
C1COCCN1C2=CC=C(C=C2)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 148°C to 152°C
Quantity 5 g
Formula Weight 208.22
Percent Purity 98%
Chemical Name or Material 4-(4-Nitrophenyl)morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'