missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Nitrobenzyloxy)benzaldehyde, 98%, Thermo Scientific™

Product Code. 11448923
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448923 5 g 5g
11458923 25 g 25g
11468923 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448923 Supplier Thermo Scientific Alfa Aesar Supplier No. A11067.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67565-48-4
Molecular Formula C14H11NO4
Molecular Weight (g/mol) 257.25
MDL Number MFCD00017041
InChI Key RIYUFYPNHRLRON-UHFFFAOYSA-N
Synonym 4-4-nitrobenzyloxy benzaldehyde, 4-4-nitrophenyl methoxy benzaldehyde, 4-4-nitrobenzyl oxy benzaldehyde, benzaldehyde,4-4-nitrophenyl methoxy, cbmicro_009110, cambridge id 5303589, 4-4-formylphenoxy methyl nitrobenzene
PubChem CID 735836
IUPAC Name 4-[(4-nitrophenyl)methoxy]benzaldehyde
SMILES [O-][N+](=O)C1=CC=C(COC2=CC=C(C=O)C=C2)C=C1

Specifications

Melting Point 128°C to 132°C
Quantity 5 g
Beilstein 1884256
Sensitivity Air Sensitive
Formula Weight 257.25
Percent Purity 98%
Chemical Name or Material 4- (4-Nitrobenzyloxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.