missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester, 97%

Product Code. 11317569
Change view
Click to view available options
Quantity:
1g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317569 1g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11317569 Supplier Alfa Aesar™ Supplier No. H28034.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 364794-79-6
Molecular Formula C17H26BNO3
MDL Number MFCD04974052
Quantity 1g
Synonym 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 4-morpholinomethyl phenylboronic acid pinacol ester, 4-4-morpholinomethyl phenylboronic acid pinacol ester, 4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 4-methylmorpholino phenylboronic acid, pinacol ester, 4-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 4-morpholinomethyl phenylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-morpholin-4-ylmethyl phenyl-1,3,2-dioxaborolane
InChI Key JOIXYIWXEYXHHG-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3CCOCC3
IUPAC Name 4-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine
Molecular Weight (g/mol) 303.209
PubChem CID 2795502
Formula Weight 303.2
Percent Purity 97%
Chemical Name or Material 4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.