missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Morpholinyl)benzonitrile, 98%, Thermo Scientific™

Product Code. 15428558
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428558 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15428558 Supplier Thermo Scientific Alfa Aesar Supplier No. H61963.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10282-31-2
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD00100115
InChI Key ZSCUWVQXQDCSRV-UHFFFAOYSA-N
PubChem CID 394808
IUPAC Name 4-morpholin-4-ylbenzonitrile
SMILES C1COCCN1C2=CC=C(C=C2)C#N

Specifications

Melting Point 85°C
Color White to Brown
Quantity 5 g
UN Number UN3439
Synonym 4-morpholinobenzonitrile, 4-4-cyanophenyl morpholine, 4-morpholin-4-yl benzonitrile, 4-4-morpholinyl benzonitrile, 4-morpholin-4-yl-benzonitrile, benzonitrile, 4-4-morpholinyl, benzonitrile,4-4-morpholinyl, 4-morpholin-4-ylbenzenecarbonitrile, pubchem18708, acmc-1bqst
Solubility Information Soluble in water.
Formula Weight 188.23
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-(4-Morpholinyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.