missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylpiperazino)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10190672
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10190672 1 g 1g
11380577 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10190672 Supplier Thermo Scientific Maybridge Supplier No. CC18704DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 27913-99-1
Molecular Formula C12H17N2O
Molecular Weight (g/mol) 205.28
MDL Number MFCD00452233
InChI Key PFODEVGLOVUVHS-UHFFFAOYSA-O
Synonym 4-4-methylpiperazin-1-yl benzaldehyde, 4-4-methylpiperazino benzaldehyde, 1-4-formylphenyl-4-methylpiperazine, 4-4-methylpiperazinyl benzaldehyde, 4-4-methylpiperazino benzenecarbaldehyde, 4-4-methyl-piperazin-1-yl-benzaldehyde, benzaldehyde, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzaldehyde, pubchem20530, acmc-1cazr
PubChem CID 736533
IUPAC Name 4-(4-methylpiperazin-1-yl)benzaldehyde
SMILES C[NH+]1CCN(CC1)C1=CC=C(C=O)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 4-(4-Methylpiperazino)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.