missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylpiperazino)aniline, 97%, Thermo Scientific™

Product Code. p-2699848
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10029631 1 g 1g
10110902 10 g 10g
10585655 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10029631 Supplier Thermo Scientific Maybridge Supplier No. SS00001DA
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 16153-81-4
Molecular Formula C11H18N3
Molecular Weight (g/mol) 192.29
MDL Number MFCD00172703
InChI Key MOZNZNKHRXRLLF-UHFFFAOYSA-O
Synonym 4-4-methylpiperazino aniline, 4-4-methylpiperazin-1-yl aniline, 4-methyl-1-4-aminophenyl piperazine, 4-4-methyl-1-piperazinyl aniline, 4-4-methylpiperazin-1-yl phenylamine, 1-4-aminophenyl-4-methylpiperazine, 4-4-methyl-piperazin-1-yl-phenylamine, 4-4-methylpiperazin-1-yl benzenamine, benzenamine, 4-4-methyl-1-piperazinyl, 4-4-methylpiperazinyl phenylamine
PubChem CID 737253
IUPAC Name 4-(4-methylpiperazin-1-yl)aniline
SMILES C[NH+]1CCN(CC1)C1=CC=C(N)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 4-(4-Methylpiperazino)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.