missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylpiperazino)acetophenone, 98%, Thermo Scientific™

Product Code. 10764081 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 10764081

Brand: Thermo Scientific Acros 152912500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 26586-55-0
CAS Min % 97.5
CAS Max % 100.0
Melting Point 96°C to 100°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C13H18N2O
MDL Number MFCD00023297
Quantity 250 mg
Synonym 1-4-4-methylpiperazino phenyl-1-ethanone, 1-4-4-methylpiperazin-1-yl phenyl ethanone, p-4-methylpiperazino-acetophenone, acetophenone, 4'-4-methyl-1-piperazinyl, 1-4-4-methylpiperazin-1-yl phenyl ethan-1-one, ethanone, 1-4-4-methyl-1-piperazinyl phenyl, ethanone,1-4-4-methyl-1-piperazinyl phenyl
InChI Key IRIZGAMYKHTLKS-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(C=C1)N2CCN(CC2)C
IUPAC Name 1-[4-(4-methylpiperazin-1-yl)phenyl]ethanone
Molecular Weight (g/mol) 218.3
PubChem CID 33551
Formula Weight 218.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Methylpiperazino)acetophenone, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.