missing translation for 'onlineSavingsMsg'
Learn More

4-(4-methylpiperazine-1-carbonyl)phenylboronic acid, 97%, Maybridge

Product Code. 10144004
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10144004 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10144004 Supplier Thermo Scientific Maybridge Supplier No. MAY00022DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 832114-06-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C18H27BN2O3
MDL Number MFCD05863914
Quantity 1 g
Synonym 4-methylpiperazin-1-yl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-methylpiperazine-1-yl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-methyl-piperazine-1-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-phenyl-methanone, 4-4-methylpiperazine-1-carbonyl phenylboronic acid, pinacol ester, 1-methyl-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, 4-4-methylpiperazine-1-carbonyl phenylboronic acid pinacol ester, 4-4-methyl-1-piperazinylcarbonyl benzeneboronic acid pinacol ester, 4-methylpiperazin-1-yl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-4-methylpiperazine-1-carbonyl phenyl boronic acid pinacol ester, 1-methyl-4-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine
InChI Key TXCDVBDHDQAWLR-UHFFFAOYSA-N
SMILES CN1CCN(CC1)C(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
IUPAC Name 1-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine
Molecular Weight (g/mol) 330.24
PubChem CID 2760030
Formula Weight 330.23
Percent Purity 97%
Chemical Name or Material 4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid, pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.