missing translation for 'onlineSavingsMsg'
Learn More

4-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde, 97%, Thermo Scientific™

Product Code. 10086252
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10086252 1 g Amber Glass Bottle 1g
10381293 250 mg Amber Glass Bottle 250mg
10634205 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10086252 Supplier Thermo Scientific Maybridge Supplier No. CC45604DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 439691-80-2
Molecular Formula C13H18N2O
Molecular Weight (g/mol) 218.3
InChI Key DJJFKXUSAXIMLS-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl methyl benzaldehyde, 1-4-formylbenzyl-4-methylpiperazine, benzaldehyde,4-4-methyl-1-piperazinyl methyl, 4-4-methylpiperazino methyl benzaldehyde, 4-4-methylpiperazinyl methyl benzaldehyde, 4-4-methyl piperazin-1-ylmethyl benzaldehyde, 4-4-methyl-1-piperazinyl methyl benzaldehyde
PubChem CID 7164651
IUPAC Name 4-[(4-methylpiperazin-1-yl)methyl]benzaldehyde
SMILES CN1CCN(CC1)CC2=CC=C(C=C2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 218.3
Percent Purity 97%
Chemical Name or Material 4-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.