missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenedianiline, 97%, Thermo Scientific™

Product Code. p-163141 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
Unit Size:
250g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10183060

Brand: Thermo Scientific Acros 158042500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-77-9
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.27
MDL Number MFCD00007914
InChI Key YBRVSVVVWCFQMG-UHFFFAOYSA-N
Synonym 4,4'-methylenedianiline, 4,4'-diaminodiphenylmethane, dadpm, methylenedianiline, dianilinomethane, tonox, p,p'-methylenedianiline, dianilinemethane, bis 4-aminophenyl methane, 4-4-aminobenzyl aniline
PubChem CID 7577
ChEBI CHEBI:32506
IUPAC Name 4-[(4-aminophenyl)methyl]aniline
SMILES NC1=CC=C(CC2=CC=C(N)C=C2)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 89°C to 94°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH2(C6H4NH2)2
Quantity 250 g
Beilstein 13, 238
Merck Index 15, 2982
Formula Weight 198.27
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4, 4'-Methylenedianiline, 97%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.