missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(phenyl isocyanate), 98%

Product Code. 11322767
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322767 500 g 500g
11302767 25 g 25g
11312767 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11322767 Supplier Thermo Scientific Alfa Aesar Supplier No. L15740.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 101-68-8 | C15H10N2O2 | 250.257 g/mol

4,4'-Methylenebis(phenyl isocyanate) acts as a monomer in the production of polyurethane. The rigid polyurethane foams are good thermal insulators, which find applications in all freezers and refrigerators, as well as in buildings. It is also used as an industrial strength adhesive, which is available to end consumers as various high-strength bottled glue preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Methylenebis(phenyl isocyanate) acts as a monomer in the production of polyurethane. The rigid polyurethane foams are good thermal insulators, which find applications in all freezers and refrigerators, as well as in buildings. It is also used as an industrial strength adhesive, which is available to end consumers as various high-strength bottled glue preparations.

Solubility
Insoluble in water. Soluble in acetone, benzene and kerosene.

Notes
Incompatible with water, amines, strong bases, alcohols and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-68-8
Molecular Formula C15H10N2O2
Molecular Weight (g/mol) 250.257
MDL Number MFCD00036131
InChI Key UPMLOUAZCHDJJD-UHFFFAOYSA-N
Synonym 4,4'-diphenylmethane diisocyanate, diphenylmethane diisocyanate, isonate, 4,4'-diisocyanatodiphenylmethane, p,p'-diphenylmethane diisocyanate, bis 4-isocyanatophenyl methane, 4,4'-methylenebis phenyl isocyanate, methylbisphenyl isocyanate, 1,1'-methylenebis 4-isocyanatobenzene, methylene diphenyl diisocyanate
PubChem CID 7570
ChEBI CHEBI:53218
IUPAC Name 1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)N=C=O)N=C=O

Specifications

Melting Point 41°C to 44°C
Density 1.239
Boiling Point 194°C (5 mmHg)
Flash Point 196°C (385°F)
Odor Weak, Characteristic
Quantity 500 g
UN Number UN2811
Beilstein 797662
Sensitivity Moisture sensitive
Solubility Information Insoluble in water. Soluble in acetone,benzene and kerosene.
Formula Weight 250.26
Percent Purity 98%
Chemical Name or Material 4,4'-Methylenebis(phenyl isocyanate)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4,4'-Methylenebis(phenyl isocyanate)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: 4,4-Methylenediphenyl diisocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.