missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(cyclohexylamine), TRC

Product Code. 30474805
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474805 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474805 Supplier Toronto Research Chemicals Supplier No. TRCM22734010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1761-71-3
Molecular Formula C13 H26 N2
Quantity 10 g
Synonym Cyclohexylamine, 4,4-methylenebis- (6CI), Cyclohexylamine, 4,4'-methylenebis- (8CI), 4,4'-Methylenebis[cyclohexanamine], 4,4'-Diaminocyclohexylmethane, 4,4'-Diaminodicyclohexylmethane, 4,4'-Methylenebis(aminocyclohexane), 4,4'-Methylenebis(cyclohexylamine), 4,4'-Methylenedicyclohexanamine, 4,4'-Methylenedicyclohexylamine, 4,4'-Methylenedicyclohexyldiamine, 4-(4-Aminocyclohexylmethyl)-1-cyclohexylamine, 4-(4-Aminocyclohexylmethyl)cyclohexanamine, Amicure PACM, Amicure PACM 20, BACM, Bis(4-aminocyclohexyl)methane, Bis(p-aminocyclohexane)methane, Bis(p-aminocyclohexyl)methane, DDCM, Di(p-aminocyclohexyl)methane, Dicyan, Dicyclohexylmethane-4,4'-diamine, HLR 4219, HLR 4448, Methylenebis(4-aminocyclohexane), Methylenebis(4-cyclohexylamine), PACM, PACM 20, Resoltech 1084, S 509361, Vestamin PACM, Wondamine HM, p,p'-Diaminodicyclohexylmethane, Cyclohexanamine, 4,4'-methylenebis-
InChI Formula InChI=1S/C13H26N2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h10-13H,1-9,14-15H2
SMILES NC1CCC(CC2CCC(N)CC2)CC1
IUPAC Name 4-[(4-aminocyclohexyl)methyl]cyclohexan-1-amine
Molecular Weight (g/mol) 210.36
Formula Weight 210.2096
Chemical Name or Material 4,4'-Methylenebis(cyclohexylamine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.