missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2-methylcyclohexylamine), 99%, mixture of isomers, Thermo Scientific Chemicals

Product Code. 15234814
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6864-37-5
Molecular Formula C15H30N2
Molecular Weight (g/mol) 238.42
MDL Number MFCD00075535
InChI Key IGSBHTZEJMPDSZ-UHFFFAOYNA-N
Synonym 4,4'-methylenebis 2-methylcyclohexanamine, bis 4-amino-3-methylcyclohexyl methane, cyclohexanamine, 4,4'-methylenebis 2-methyl, 4,4'-methylenebis 2-methylcyclohexylamine, 3,3'-dimethyl-4,4'-diaminodicyclohexylmethane, 2,2'-dimethyl-4,4'-methylenebis cyclohexylamine, 4-4-amino-3-methylcyclohexyl methyl-2-methylcyclohexan-1-amine, 4,4'-methylenebis 2-methylcyclohexyl-amine, 2,2'-dimethyl-4,4'methylenebis cyclohexylamine
PubChem CID 91555
IUPAC Name 4-[(4-amino-3-methylcyclohexyl)methyl]-2-methylcyclohexan-1-amine
SMILES CC1CC(CC2CCC(N)C(C)C2)CCC1N

Specifications

Density 0.9400g/mL
Boiling Point 93.0°C to 100.0°C
Flash Point 113°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 250 mL
Specific Gravity 0.94
Viscosity 142 mPa.s (20°C)
Formula Weight 238.41
Percent Purity 99%
Chemical Name or Material 4, 4'-Methylenebis(2-methylcyclohexylamine)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4,4'-Methylenebis(2-methylcyclohexylamine)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'