missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2-chloroaniline) (~90%), TRC

Product Code. 30483099
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483099 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483099 Supplier Toronto Research Chemicals Supplier No. TRCM3042805G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-Methylene-bis(2-chloroaniline)
CAS 101-14-4
Molecular Formula C13 H12 Cl2 N2
Quantity 5 g
Synonym Benzenamine','4,4′-methylenebis[2-chloro- (9CI','ACI), 4,4′-Methylenebis[2-chlorobenzenamine] (ACI), Aniline','4,4′-methylenebis[2-chloro- (6CI','8CI), 2,2′-Dichloro-4,4′-methylenedianiline, 3,3′-Dichloro-4,4′-diaminodiphenylmethane, 4,4′-Methylenebis(2-chloraniline), 4,4′-Methylenebis[2-chloroaniline], 4,4′-Methylenebis[o-chloroaniline], [4-(4-Amino-3-chlorobenzyl)-2-chlorophenyl]amine, Bis(3-chloro-4-aminophenyl)methane, Bis(4-amino-3-chlorophenyl)methane, Bisamine A, Bisamine S, CPP 100, Cuamine K, Cuamine M, Cuamine MT, Curalon M, Curene 442, Diamet Kh, Diamet X, DKh, HR 100, HR 100 (amine), Ihara Cuamine MT, Isocross SM, Kuralon, MBOCA, Methylenebis(3-chloro-4-aminobenzene), Methylenebis(chloraniline), Methylenebis(o-chloroaniline), Millionate M, MOCA, MOCA (curing agent), NSC 52954, Pandex E, Pandex M 3202, Quodorole, SL 4037, Vibracure 133, Vibracure A 133
InChI Formula InChI=1S/C13H12Cl2N2/c14-10-6-8(1-3-12(10)16)5-9-2-4-13(17)11(15)7-9/h1-4,6-7H,5,16-17H2
SMILES Nc1ccc(Cc2ccc(N)c(Cl)c2)cc1Cl
IUPAC Name 4-[(4-amino-3-chlorophenyl)methyl]-2-chloroaniline
Molecular Weight (g/mol) 267.15
Formula Weight 266.0378
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.