missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxyphenyl)pyrimidine-2-thiol, ≥95%, Thermo Scientific™

Product Code. p-2700683
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10573202 1 g 1g
10130832 10 g 10g
10071724 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10573202 Supplier Thermo Scientific Maybridge Supplier No. KM04163DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175202-77-4
Molecular Formula C11H10N2OS
Molecular Weight (g/mol) 218.274
MDL Number MFCD00085109
InChI Key KSXDNCUHISGCTF-UHFFFAOYSA-N
Synonym 4-4-methoxyphenyl pyrimidine-2-thiol, 4-4-methoxyphenyl-3h-pyrimidine-2-thione, maybridge1_004759, 4-4-methoxyphenyl-2-pyrimidinethiol, 4-4-methoxy-phenyl-pyrimidine-2-thiol, 6-4-methoxyphenyl-1h-pyrimidine-2-thione, 4-4-methoxyphenyl-2-pyrimidinyl hydrosulfide
PubChem CID 2736768
IUPAC Name 6-(4-methoxyphenyl)-1H-pyrimidine-2-thione
SMILES COC1=CC=C(C=C1)C2=CC=NC(=S)N2

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 218.28
Percent Purity 95%
Chemical Name or Material 4-(4-Methoxyphenyl)pyrimidine-2-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.