missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxyphenyl)butyric Acid, TRC

Product Code. 30481234
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481234 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481234 Supplier Toronto Research Chemicals Supplier No. TRCM3295681G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(4-Methoxyphenyl)butyric Acid
CAS 4521-28-2
Molecular Formula C11H14O3
Quantity 1 g
Synonym 4-Methoxybenzenebutanoic Acid, 4-(4-Methoxyphenyl)butanoic Acid, 4-(p-Methoxyphenyl)butyric Acid, NSC 407553, gamma-(p-Methoxyphenyl)-butyric Acid
InChI Formula InChI=1S/C11H14O3/c1-14-10-7-5-9(6-8-10)3-2-4-11(12)13/h5-8H,2-4H2,1H3,(H,12,13)
SMILES COc1ccc(CCCC(=O)O)cc1
IUPAC Name 4-(4-methoxyphenyl)butanoic acid
Molecular Weight (g/mol) 194.23
Formula Weight 194.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.