missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Isocyanatopyrid-2-yl)morpholine, ≥90%, Thermo Scientific™

Product Code. 10642855
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10642855 250 mg Amber Glass Bottle 250mg
10406613 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10642855 Supplier Thermo Scientific Maybridge Supplier No. CC43106CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 876316-43-7
Molecular Formula C10H11N3O2
Molecular Weight (g/mol) 205.22
MDL Number MFCD08271914
InChI Key LRVCWTODNVVIOK-UHFFFAOYSA-N
Synonym 4-4-isocyanatopyrid-2-yl morpholine, 4-4-isocyanatopyridin-2-yl morpholine, morpholine,4-4-isocyanato-2-pyridinyl, 2-morpholin-4-ylpyridin-4-isocyanate, 4-4-isocyanatopyrid-2-yl-morpholine, 2-morpholin-4-yl pyridin-4-yl isocyanate, 4-isocyanato-2-morpholin-4-yl pyridine
PubChem CID 18525762
IUPAC Name 4-(4-isocyanatopyridin-2-yl)morpholine
SMILES O=C=NC1=CC(=NC=C1)N1CCOCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 205.22
Percent Purity 95%
Chemical Name or Material 4-(4-Isocyanatopyrid-2-yl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.