missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Iodophenyl)morpholine, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Synonym | 4-4-iodophenyl morpholine, 4-iodophenylmorpholine, morpholine,4-4-iodophenyl, morpholine, 4-4-iodophenyl, 1-iodo-4-morpholin-4-yl benzene |
Formula Weight | 289.11 |
Percent Purity | 97% |
Chemical Name or Material | 4-(4-Iodophenyl)morpholine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'