missing translation for 'onlineSavingsMsg'
Learn More

4-(4’-Hydroxyphenyl)-2-butanone, TRC

Product Code. 30133921
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133921 100 mg 100mg
30004080 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133921 Supplier Toronto Research Chemicals Supplier No. TRCH949105100MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5471-51-2
Molecular Formula C10 H12 O2
Molecular Weight (g/mol) 164.2
Synonym Frambinone, 4-(4-Hydroxyphenyl)butan-2-one, 2-Butanone, 4-(4-hydroxyphenyl)-, 2-Butanone, 4-(p-hydroxyphenyl)- (6CI,7CI,8CI), 4-(4-Hydroxyphenyl)-2-butanone, (4-Hydroxybenzyl)acetone, (p-Hydroxybenzyl)acetone, 1-(4-Hydroxyphenyl)-3-butanone, 1-(p-Hydroxyphenyl)-3-butanone, 4-(3-Oxobutyl)phenol, 4-(4-Hydroxy-1-phenyl)-2-butanone, 4-(4'-Hydroxyphenyl)-2-butanone, 4-(p-Hydroxyphenyl)-2-butanone, Frambinone, NSC 26515, Oxyphenylon, Raspberry ketone, Rheosmin
IUPAC Name 4-(4-hydroxyphenyl)butan-2-one
SMILES CC(=O)CCc1ccc(O)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H12O2/c1-8(11)2-3-9-4-6-10(12)7-5-9/h4-7,12H,2-3H2,1H3
Formula Weight 164.0837
Percent Purity >95%
Chemical Name or Material 4-Hydroxyphenyl-2-Butanone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.