missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenyl)-2-butanone, 99+%

Product Code. 10249633 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10249633 1 kg Glass bottle 1kg
10449490 50 g Glass bottle 50g
10614682 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10249633 Supplier Thermo Scientific Acros Supplier No. 176470010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5471-51-2 | C10H12O2 | 164.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5471-51-2
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00002394
InChI Key NJGBTKGETPDVIK-UHFFFAOYSA-N
Synonym 4-4-hydroxyphenyl-2-butanone, raspberry ketone, 4-4-hydroxyphenyl butan-2-one, frambinone, oxyphenalon, rheosmin, 4-p-hydroxyphenyl-2-butanone, 4-hydroxybenzylacetone, betuligenol, rasketone
PubChem CID 21648
ChEBI CHEBI:68656
IUPAC Name 4-(4-hydroxyphenyl)butan-2-one
SMILES CC(=O)CCC1=CC=C(C=C1)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 82°C to 84°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H4CH2CH2COCH3
Quantity 1 kg
Formula Weight 164.2
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-(4-Hydroxyphenyl)-2-butanone, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 4-(4-Hydroxyphenyl)-2-butanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.