missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodophenyl]acetic Acid (Triiodothyroacetic Acid), Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
[4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodophenyl]acetic Acid (Triiodothyroacetic Acid)
Specifications
Specifications
| Chemical Name or Material | 3,3,5-Triiodo Thyroacetic Acid |
| CAS | 51-24-1 |
| Analyte or Component Names | 3,3,5-Triiodo Thyroacetic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H9 I3 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Levothyroxine Sodium Imp. C (EP), Levothyroxine Imp. C (EP), Liothyronine Sodium Imp. C (EP), Liothyronine Imp. C (EP), Benzeneacetic acid, 4-(4-hydroxy-3-iodophenoxy)-3,5-diiodo-, Acetic acid, [4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]- (8CI), Thyroacetic acid, 3,3',5-triiodo- (6CI), 3,3',5-Triiodothyroa, Tiratricol, Levothyroxine Sodium Impurity C, Liothyronine Sodium Impurity C, Levothyroxine Impurity C, Liothyronine Impurity C |
| InChI Formula | InChI=1S/C14H9I3O4/c15-9-6-8(1-2-12(9)18)21-14-10(16)3-7(4-11(14)17)5-13(19)20/h1-4,6,18H,5H2,(H,19,20) |
| SMILES | OC(=O)Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1 |
| IUPAC Name | 2-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]acetic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction