missing translation for 'onlineSavingsMsg'
Learn More

4,4'-(Hexafluoroisopropylidene)dianiline, 98%, Thermo Scientific Chemicals

Product Code. 10473015
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1095-78-9
Molecular Formula C15H12F6N2
Molecular Weight (g/mol) 334.27
MDL Number MFCD00039146
InChI Key BEKFRNOZJSYWKZ-UHFFFAOYSA-N
Synonym 2,2-bis 4-aminophenyl hexafluoropropane, 4,4'-perfluoropropane-2,2-diyl dianiline, 4,4'-hexafluoroisopropylidene dianiline, 4-2-4-aminophenyl-1,1,1,3,3,3-hexafluoropropan-2-yl aniline, benzenamine, 4,4'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, acmc-1bon7, bekfrnozjsywkz-uhfffaoysa, 2,2-bis-4-aminophenyl hexafluoropropane, 4,4'-1,1,1,3,3,3-hexafluoropropane-2,2-diyl dianiline
PubChem CID 622236
IUPAC Name 4-[2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]aniline
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)N)(C(F)(F)F)C(F)(F)F)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 195.0°C to 199.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CF3)2C(C6H4NH2)2
Quantity 5 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 334.27
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-(Hexafluoroisopropylidene)dianiline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'