missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)butyric acid, 97%

Product Code. 15473347
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473347 250 mg 250mg
11321969 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473347 Supplier Thermo Scientific Alfa Aesar Supplier No. H26035.MD
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 589-06-0 | C10H11FO2 | 182.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-06-0
Molecular Formula C10H11FO2
Molecular Weight (g/mol) 182.19
MDL Number MFCD03788503
InChI Key XVQYBBYOYJXQBF-UHFFFAOYSA-N
Synonym 4-4-fluorophenyl butanoic acid, 4-4-fluorophenyl butyric acid, 4-4-fluoro-phenyl-butyric acid, 4-p-fluorophenyl butyric acid, benzenebutanoic acid, 4-fluoro, 4-4-fluorophenyl butanoicacid, acmc-1awzu, 4-fluorobenzenebutanoic acid, 4-4-fluorophenyl butyricacid, benzenebutanoic acid,4-fluoro
PubChem CID 68524
IUPAC Name 4-(4-fluorophenyl)butanoic acid
SMILES OC(=O)CCCC1=CC=C(F)C=C1

Specifications

Melting Point 44°C to 46°C
Quantity 250 mg
Formula Weight 182.2
Percent Purity 97%
Chemical Name or Material 4-(4-Fluorophenyl)butyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.