missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridin-2(1H)-one, TRC

Product Code. 30401946
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401946 250 mg -
30401836 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401946 Supplier Toronto Research Chemicals Supplier No. TRCF595210250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132812-72-7
Molecular Formula C17 H18 F N O
Molecular Weight (g/mol) 271.33
IUPAC Name 4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydro-1H-cycloocta[b]pyridin-2-one
SMILES Fc1ccc(cc1)C2=CC(=O)NC3=C2CCCCCC3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H18FNO/c18-13-9-7-12(8-10-13)15-11-17(20)19-16-6-4-2-1-3-5-14(15)16/h7-11H,1-6H2,(H,19,20)
Formula Weight 271.1372
Chemical Name or Material 4-(4-Fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridin-2(1H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.