missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)-4-oxobutanoic Acid, TRC

Product Code. 30398545
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398545 2.5 g 2.5g
30402414 5 g -
30401023 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398545 Supplier Toronto Research Chemicals Supplier No. TRCF6322832.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-77-8
Molecular Formula C10 H9 F O3
Molecular Weight (g/mol) 196.175
Synonym Benzenebutanoic acid, 4-fluoro-γ-oxo- (9CI, ACI), 4-Fluoro-γ-oxobenzenebutanoic acid (ACI), Propionic acid, 3-(p-fluorobenzoyl)- (7CI, 8CI), 3-(4-Fluorobenzoyl)propanoic acid, 3-(4-Fluorobenzoyl)propionic acid, 3-(p-Fluorobenzoyl)propionic acid, 4-(4-Fluorophenyl)-4-oxobutanoic acid, 4-(4-Fluorophenyl)-4-oxobutyric acid, 4-Fluorobenzoylpropionic acid, 4-Oxo-4-(p-fluorophenyl)butyric acid, Ba 2833, NSC 408180, β-(4-Fluorobenzoyl)propionic acid, β-(p-Fluorobenzoyl)propionic acid, γ-(4-Fluorophenyl)-γ-oxobutanoic acid
IUPAC Name 4-(4-fluorophenyl)-4-oxobutanoic acid
SMILES OC(=O)CCC(=O)c1ccc(F)cc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C10H9FO3/c11-8-3-1-7(2-4-8)9(12)5-6-10(13)14/h1-4H,5-6H2,(H,13,14)
Formula Weight 196.054
Chemical Name or Material 4-(4-Fluorophenyl)-4-oxobutanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.