missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)-1,3-thiazole, TRC

Product Code. 30398035
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398035 10 mg
30395386 100 mg
30406072 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398035 Supplier Toronto Research Chemicals Supplier No. TRCF40257810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 383145-29-7
Molecular Formula C9H6NFS
Molecular Weight (g/mol) 179.21
SMILES Fc1ccc(cc1)c2cscn2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H6FNS/c10-8-3-1-7(2-4-8)9-5-12-6-11-9/h1-6H
Formula Weight 179.0205
Chemical Name or Material 4-(4-Fluorophenyl)-1,3-thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.