missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenoxy)phenylacetic acid, 97%, Thermo Scientific™

Product Code. 15445787
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15445787

Brand: Thermo Scientific Alfa Aesar H32998.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 41073-15-8
Melting Point 98°C to 100°C
Molecular Formula C14H11FO3
MDL Number MFCD08446956
Quantity 1 g
UN Number UN3077
Sensitivity Light and moisture sensitive
Synonym 4-4-fluorophenoxy phenylacetic acid, 2-4-4-fluorophenoxy phenyl acetic acid, 4-4-fluorophenoxy phenyl acetic acid, acmc-20amw0
InChI Key VMMSFFIABDLZIP-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CC(=O)O)OC2=CC=C(C=C2)F
IUPAC Name 2-[4-(4-fluorophenoxy)phenyl]acetic acid
Molecular Weight (g/mol) 246.237
PubChem CID 15549820
Formula Weight 246.24
Percent Purity 97%
Chemical Name or Material 4-(4-Fluorophenoxy)phenylacetic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.