missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorobenzoyl)pyridine, 99%

Product Code. 11496026
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11496026

Brand: Thermo Scientific Alfa Aesar B22327.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41538-36-7 | C12H8FNO | 201.20 g/mol

4-(4-Fluorobenzoyl)pyridine is used for increasing cardiac contractility and for the treatment of congestive heart failure, and pharmaceutical compositions and for the preparation thereof.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Fluorobenzoyl)pyridine is used for increasing cardiac contractility and for the treatment of congestive heart failure, and pharmaceutical compositions and for the preparation thereof.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41538-36-7
Molecular Formula C12H8FNO
Molecular Weight (g/mol) 201.20
MDL Number MFCD02683088
InChI Key WTRWBYGUMQEFFI-UHFFFAOYSA-N
Synonym 4-4-fluorobenzoyl pyridine, 4-fluorophenyl pyridin-4-yl methanone, methanone, 4-fluorophenyl-4-pyridinyl, 4-fluorophenyl-pyridin-4-ylmethanone, 4-fluorophenyl 4-pyridyl ketone, acmc-20amw5, d0j0wo, 4-pyridyl p-fluorophenyl ketone, 4-4-fluorophenyl carbonyl pyridine, 4-fluorophenyl 4-pyridinyl methanone
PubChem CID 7023019
SMILES FC1=CC=C(C=C1)C(=O)C1=CC=NC=C1

Specifications

Melting Point 85°C to 87°C
Quantity 1 g
IUPAC Name 4-(4-fluorobenzoyl)pyridine
Formula Weight 201.2
Percent Purity 99%
Chemical Name or Material 4-(4-Fluorobenzoyl)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.