missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%

Product Code. 11456366
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456366 5 g 5g
11446366 1 g 1g
11466366 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456366 Supplier Thermo Scientific Alfa Aesar Supplier No. B23142.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25519-78-2 | C12H15ClFNO | 243.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25519-78-2
Molecular Formula C12H15ClFNO
Molecular Weight (g/mol) 243.71
MDL Number MFCD00044912
InChI Key GPKDBZQZPNOBGM-UHFFFAOYSA-N
Synonym 4-4-fluorobenzoyl piperidine hydrochloride, 4-fluorophenyl piperidin-4-yl methanone hydrochloride, 4-fluorophenyl 4-piperidinyl ketone hydrochloride, p-fluorophenyl 4-piperidyl ketone hydrochloride, 4-fluorophenyl 4-piperidyl methanone hydrochloride, 4-fluoro-phenyl-piperidin-4-yl-methanone hydrochloride, 4-4-fluorobenzoyl piperidine hcl, 4-p-fluorobenzoyl piperidine hydrochloride, 4-4'-fluorobenzoyl piperidine hydrochloride, methanone, 4-fluorophenyl-4-piperidinyl-, hydrochloride
PubChem CID 3084438
IUPAC Name (4-fluorophenyl)-piperidin-4-ylmethanone;hydrochloride
SMILES [H+].[Cl-].FC1=CC=C(C=C1)C(=O)C1CCNCC1

Specifications

Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 243.71
Percent Purity 98%
Chemical Name or Material 4-(4-Fluorobenzoyl)piperidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.