missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dipyridyl, 98%, anhydrous, ACROS Organics™

Brand:  Acros Organics 276200050

Code : 29

Additional Details : CAS Number : 553-26-4 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10067632

  • £18.69 / 5g

In stock, ready to ship: 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

553-26-4
156.19
4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridyl, 4-4-pyridyl pyridine, 4,4'-dipyridine, 4,4-bipyridyl, 4,4-dipyridyl, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
CHEBI:30985
C1=CN=CC=C1C2=CC=NC=C2
C10H8N2
MWVTWFVJZLCBMC-UHFFFAOYSA-N
11107
4-pyridin-4-ylpyridine

Specifications

4, 4'-Dipyridyl
Glass bottle
1% Max. (K.F.)
White to Beige
5g
98%
(C5H4N)-(C5H4N)
16,25
4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridyl, 4-4-pyridyl pyridine, 4,4'-dipyridine, 4,4-bipyridyl, 4,4-dipyridyl, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
MWVTWFVJZLCBMC-UHFFFAOYSA-N
4-pyridin-4-ylpyridine
11107
156.19
98%
Authentic
0.1% max.
305.0°C
110.0°C to 114.0°C
553-26-4
C10H8N2
23,2
15,3389
Solubility in water: 4,5g/L (25°C). Other solubilities: freely soluble in alcohol, benzene, chloroform,, ether,
C1=CN=CC=C1C2=CC=NC=C2
156.19
CHEBI:30985
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4,4`-Dipyridyl, anhydrous

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS Documents