missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyldihydrofuran-2,3-dione, TRC

Product Code. 30385256
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385256 50 mg
30371343 500 mg
30369717 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385256 Supplier Toronto Research Chemicals Supplier No. TRCD47694350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13031-04-4
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
Synonym Dexpanthenol Impurity 5
IUPAC Name 4,4-dimethyloxolane-2,3-dione
SMILES CC1(C)COC(=O)C1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H8O3/c1-6(2)3-9-5(8)4(6)7/h3H2,1-2H3
Formula Weight 128.0473
Chemical Name or Material 4,4-Dimethyldihydrofuran-2,3-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.