missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethylcyclohexan-1-ol, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 932-01-4 |
---|---|
Molecular Formula | C8H16O |
Molecular Weight (g/mol) | 128.22 |
MDL Number | MFCD00101954 |
InChI Key | VUQOIZPFYIVUKD-UHFFFAOYSA-N |
PubChem CID | 136735 |
IUPAC Name | 4,4-dimethylcyclohexan-1-ol |
SMILES | CC1(C)CCC(O)CC1 |
Description
Chemical Identifiers
932-01-4 | |
128.22 | |
VUQOIZPFYIVUKD-UHFFFAOYSA-N | |
4,4-dimethylcyclohexan-1-ol |
C8H16O | |
MFCD00101954 | |
136735 | |
CC1(C)CCC(O)CC1 |
Specifications
932-01-4 | |
100.0 | |
C8H16O | |
4,4-dimethylcyclohexanol, cyclohexanol, 4,4-dimethyl, 4,4-dimethyl-cyclohexanol, cyclohexanol,4,4-dimethyl, 4,4-dimethyl-1-cyclohexanol, 4,4-dimethyl cyclohexan-1-ol | |
CC1(C)CCC(O)CC1 | |
128.22 | |
128.21 | |
4, 4-Dimethylcyclohexan-1-ol |
94.5 | |
95% | |
MFCD00101954 | |
VUQOIZPFYIVUKD-UHFFFAOYSA-N | |
4,4-dimethylcyclohexan-1-ol | |
136735 | |
95% |
Safety and Handling
GHS P Statement Causes serious eye irritation. Causes skin irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin
Warning