missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl Retinoic Acid, TRC

Product Code. 30132957
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
10 mg
Unit Size:
0.5mg
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132957 10 mg 10mg
30012285 2.5 mg 2.5mg
30004138 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132957 Supplier Toronto Research Chemicals Supplier No. TRCD47920010MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104182-09-4
Molecular Formula C22 H32 O2
Molecular Weight (g/mol) 328.49
Synonym SRI 5397-41X
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,3,3,6,6-pentamethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
SMILES C\C(=C/C=C/C(=C/C(=O)O)/C)\C=C\C1=C(C)C(C)(C)CCC1(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H32O2/c1-16(9-8-10-17(2)15-20(23)24)11-12-19-18(3)21(4,5)13-14-22(19,6)7/h8-12,15H,13-14H2,1-7H3,(H,23,24)/b10-8+,12-11+,16-9+,17-15+
Formula Weight 328.2402
Chemical Name or Material 4,4-Dimethyl Retinoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.