missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-cyclohexen-1-one, 97%, Thermo Scientific Chemicals

Catalog Number 10471501
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1073-13-8
C8H12O
124.18
MFCD00009695
HAUNPYVLVAIUOO-UHFFFAOYSA-N
4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one
136839
4,4-dimethylcyclohex-2-en-1-one
CC1(C)CCC(=O)C=C1
This item is not returnable. View return policy
1073-13-8
C8H12O
124.18
MFCD00009695
HAUNPYVLVAIUOO-UHFFFAOYSA-N
4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one
136839
4,4-dimethylcyclohex-2-en-1-one
CC1(C)CCC(=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Brown
Density 0.9400g/mL
Boiling Point 72.0°C to 73.5°C (20.0 mmHg)
Flash Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C6H6(=O)
Refractive Index 1.4710 to 1.4740
Quantity 5 g
Specific Gravity 0.94
Formula Weight 124.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4, 4-Dimethyl-2-cyclohexen-1-one
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'