missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-cyclohexen-1-one, 96%

Product Code. p-7049645
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11450490

Brand: Thermo Scientific Alfa Aesar 017162.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1073-13-8 | C8H12O | 124.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073-13-8
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
MDL Number MFCD00009695
InChI Key HAUNPYVLVAIUOO-UHFFFAOYSA-N
Synonym 4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one
PubChem CID 136839
IUPAC Name 4,4-dimethylcyclohex-2-en-1-one
SMILES CC1(C)CCC(=O)C=C1

Specifications

Density 0.944 g/mL
Boiling Point 73°C to 74°C (14 mmHg)
Flash Point 64°C (148°F)
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 124.18
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 4,4-Dimethyl-2-cyclohexen-1-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.