missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethyl-2,2'-bipyridine, TRC

Product Code. 30347331
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347331 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347331 Supplier Toronto Research Chemicals Supplier No. TRCB429933500MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-Dimethyl-2,2'-bipyridine
CAS 1134-35-6
Molecular Formula C12 H12 N2
Quantity 500 mg
InChI Formula InChI=1S/C12H12N2/c1-9-3-5-13-11(7-9)12-8-10(2)4-6-14-12/h3-8H,1-2H3
SMILES Cc1ccnc(c1)c2cc(C)ccn2
IUPAC Name 4-methyl-2-(4-methylpyridin-2-yl)pyridine
Molecular Weight (g/mol) 184.237
Formula Weight 184.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.