missing translation for 'onlineSavingsMsg'
Learn More

4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione, TRC

Product Code. 30385688
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385688 100 mg -
30378543 50 mg 50mg
30387701 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385688 Supplier Toronto Research Chemicals Supplier No. TRCD457978100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5488-36-8
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
SMILES CC1(C)C(=O)NC(=O)c2ccccc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H11NO2/c1-11(2)8-6-4-3-5-7(8)9(13)12-10(11)14/h3-6H,1-2H3,(H,12,13,14)
Formula Weight 189.079
Chemical Name or Material 4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline-1,3-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.