missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Dimethoxytrityl chloride, 97%, Thermo Scientific Chemicals
15.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4,4'-Dimethoxytrityl chloride is used as a protective reagent for oligonucleotide synthesis. It is used for selective protection and deprotection procedures for thiol and hydroxy groups in nucleoside derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Dimethoxytrityl chloride is used as a protective reagent for oligonucleotide synthesis. It is used for selective protection and deprotection procedures for thiol and hydroxy groups in nucleoside derivatives.
Solubility
Soluble in chloroform and methanol. Partially soluble in water
Notes
Moisture sensitive, hygroscopic. Incompatible with strong oxidizing agents.
4,4′-Dimethoxytrityl chloride is used as a protective reagent for oligonucleotide synthesis. It is used for selective protection and deprotection procedures for thiol and hydroxy groups in nucleoside derivatives.
Solubility
Soluble in chloroform and methanol. Partially soluble in water
Notes
Moisture sensitive, hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 121°C to 127°C |
Quantity | 5 g |
Beilstein | 2471942 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in chloroform and methanol. Partially soluble in water |
Formula Weight | 338.83 |
Percent Purity | 98% |
Chemical Name or Material | 4,4'-Dimethoxytrityl chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'