missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxybenzil, MedChemExpress

Product Code. 16461019
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16461019 Supplier Medchem Express Supplier No. HY103610

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 4,4'-Dimethoxybenzil is a human intestinal carboxyl esterase (hiCE) inhibitor with Ki of 70 nM.

4,4'-Dimethoxybenzil is a human intestinal carboxyl esterase (hiCE) inhibitor with Ki of 70 nM.

Specifications

Chemical Name or Material 4,4'-Dimethoxybenzil
CAS 1226-42-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H14O4
Quantity 1 g
Synonym p-Anisil
Solubility Information DMSO : 50 mg/mL (184.99 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC=C(OC)C=C1)C(C2=CC=C(OC)C=C2)=O
Molecular Weight (g/mol) 270.28
Formula Weight 270.28
Percent Purity 99.3%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.