missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxybenzil, 99+%, ACROS Organics™

Brand:  Acros Organics 170450050

Code : 29

Additional Details : CAS Number : 1226-42-2 Weight : 0.05500kg

Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10012120

  • £22.70 / 5g


Estimated Shipment
17-04-2019
Add to basket

Specifications

Chemical Identifiers

1226-42-2
270.284
YNANGXWUZWWFKX-UHFFFAOYSA-N
71043
COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC
C16H14O4
MFCD00008405
4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione
1,2-bis(4-methoxyphenyl)ethane-1,2-dione

Specifications

4, 4'-Dimethoxybenzil
Glass bottle
131.0°C to 134.0°C
99+%
99.0
99+%
CH3OC6H4COCOC6H4OCH3
08, 428
YNANGXWUZWWFKX-UHFFFAOYSA-N
1,2-bis(4-methoxyphenyl)ethane-1,2-dione
71043
Fine Crystalline Powder
Authentic
Yellow
5g
1226-42-2
100.0
C16H14O4
MFCD00008405
4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione
COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC
270.284
270.27
≥99%
Videos
SDS