missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxybenzhydrol, 98+%

Product Code. 11442038
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442038 50 g 50g
11432038 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11442038 Supplier Thermo Scientific Alfa Aesar Supplier No. A14604.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 728-87-0 | C15H16O3 | 244.29 g/mol

4,4?-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.

Solubility
Soluble in water (0.44 g/L @ 25°C), methanol (0.1 g/mL).

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 728-87-0
Molecular Formula C15H16O3
Molecular Weight (g/mol) 244.29
MDL Number MFCD00008410
InChI Key ZODAOVNETBTTJX-UHFFFAOYSA-N
Synonym 4,4'-dimethoxybenzhydrol, bis 4-methoxyphenyl methanol, p,p'-dimethoxybenzhydryl alcohol, p,p'-dimethoxybenzhydrol, bis p-methoxyphenyl carbinol, benzenemethanol, 4-methoxy-.alpha.-4-methoxyphenyl, 4-methoxy-4'-methoxybenzhydrol, bis 4-methoxyphenyl methan-1-ol, benzhydrol, 4,4'-dimethoxy, acmc-209ooh
PubChem CID 69768
IUPAC Name bis(4-methoxyphenyl)methanol
SMILES COC1=CC=C(C=C1)C(O)C1=CC=C(OC)C=C1

Specifications

Melting Point 69°C to 71°C
Quantity 50 g
Beilstein 748532
Solubility Information Soluble in water (0.44g/L @ 25°C),methanol (0.1g/mL).
Formula Weight 244.29
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dimethoxybenzhydrol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.