missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4-Dimethoxy-N,N-dimethylbutan-1-amine (4-(Dimethylamino)butyraldehyde Dimethyl Acetal), Mikromol™
Description
4,4-Dimethoxy-N,N-dimethylbutan-1-amine (4-(Dimethylamino)butyraldehyde Dimethyl Acetal)
Specifications
Specifications
| Chemical Name or Material | 4-(N,N-Dimethylamino)butanal Dimethyl Acetal |
| CAS | 19718-92-4 |
| Analyte or Component Names | 4-(N,N-Dimethylamino)butanal Dimethyl Acetal |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8H19NO2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-(Dimethylamino)butyraldehyde Dimethyl Acetal, 4,4-Dimethoxy-N,N-dimethylbutanamine, 4,4-Dimethoxy-N,N-dimethylbutan-1-amine |
| SMILES | COC(CCCN(C)C)OC |
| Molecular Weight (g/mol) | 161.24 |
| Quantity | 100 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?