missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diiodobiphenyl, 99%

Product Code. p-3586359 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10124703

Brand: Thermo Scientific Acros 115120050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3001-15-8 | C12H8I2 | 406 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3001-15-8
Molecular Formula C12H8I2
Molecular Weight (g/mol) 406
MDL Number MFCD00001057
InChI Key GPYDMVZCPRONLW-UHFFFAOYSA-N
Synonym 4,4'-diiodobiphenyl, 4,4'-diiodo-1,1'-biphenyl, 4,4'-diiododiphenyl, 1,1'-biphenyl, 4,4'-diiodo, 1-iodo-4-4-iodophenyl benzene, 4,4-diiodobiphenyl, 4-4'-iodophenyl iodobenzene, 4,4;-diiodobiphenyl, 4,4'-diiododophenyl, pract, 4-iodo-1-4-iodophenyl benzene
PubChem CID 76348
IUPAC Name 1-iodo-4-(4-iodophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)I

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 201°C to 204°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula IC6H4C6H4I
Quantity 5 g
Beilstein 05, 581
Formula Weight 406
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4, 4'-Diiodobiphenyl
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Diiodobiphenyl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.