missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxybenzophenone, 98+%

Product Code. p-7022362
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11427407

Brand: Thermo Scientific Alfa Aesar A15097.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-99-4 | C13H10O3 | 214.22 g/mol

4,4'-Dihydroxybenzophenone is as a UV light stabilizer. It and its derivatives are found in cosmetics, plastics, films, adhesives and coatings, optical fiber, and printed circuit boards. It is the precursor to certain polycarbonate polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dihydroxybenzophenone is as a UV light stabilizer. It and its derivatives are found in cosmetics, plastics, films, adhesives and coatings, optical fiber, and printed circuit boards. It is the precursor to certain polycarbonate polymers.

Solubility
Soluble in water(0.45g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-99-4
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002358
InChI Key RXNYJUSEXLAVNQ-UHFFFAOYSA-N
Synonym 4,4'-dihydroxybenzophenone, bis 4-hydroxyphenyl methanone, hbp ketone, methanone, bis 4-hydroxyphenyl, bis 4-hydroxyphenyl ketone, 4,4'-dihydoxy-benzophenone, bis p-hydroxy benzophenone, p,p'-dihydroxybenzophenone, 4,4'-dihydroxydiphenyl ketone, benzophenone, 4,4'-dihydroxy
PubChem CID 69150
ChEBI CHEBI:34365
IUPAC Name bis(4-hydroxyphenyl)methanone
SMILES C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)O)O

Specifications

Melting Point 221°C to 225°C
Quantity 5 g
Beilstein 1874572
Solubility Information Soluble in water(0.45g/L).
Formula Weight 214.22
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dihydroxybenzophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.