missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluoropiperidine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11308956
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308956 1 g 1g
11318956 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308956 Supplier Thermo Scientific Alfa Aesar Supplier No. L19555.03
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 144230-52-4 | Molecular Formula: C5H10ClF2N | Molecular Weight: 157.589 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144230-52-4
Molecular Formula C5H10ClF2N
Molecular Weight (g/mol) 157.589
MDL Number MFCD03095381
InChI Key OABUKBBBSMNNPM-UHFFFAOYSA-N
Synonym 4,4-difluoropiperidine hydrochloride, 4,4-difluoropiperidine hcl, piperidine, 4,4-difluoro-, hydrochloride, 4,4-difluoropiperidine, chloride, 4,4-difluoropiperidin-1-ium chloride, 4,4-difluoropiperidinehydrochloride, pubchem6797, acmc-1bwk7, 4,4-difluoropiperidinehcl, 4,4-difluoropiperidin hcl
PubChem CID 2758351
IUPAC Name 4,4-difluoropiperidine;hydrochloride
SMILES C1CNCCC1(F)F.Cl

Specifications

Melting Point ∼172°C
Quantity 1 g
Beilstein 6812163
Sensitivity Moisture sensitive
Formula Weight 157.59
Percent Purity 98%
Chemical Name or Material 4,4-Difluoropiperidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.