missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobenzhydryl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7081303
Click to view available options
Quantity:
5 g
10 g
50 g
This item is not returnable. View return policy
27064-94-4
C13H9ClF2
238.662
MFCD00044329
FHPNLCLHMNPLEW-UHFFFAOYSA-N
4,4'-difluorobenzhydryl chloride, 4,4'-chloromethylene bis fluorobenzene, 1,1'-chloromethylene bis 4-fluorobenzene, chlorobis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethylchloride, chloro bis-4-fluorophenyl methane, bis 4-fluorophenyl chloromethane, 4,4'-difluorobenzhydrylchloride, 1-chloro 4-fluorophenyl methyl-4-fluorobenzene, 1-chloro-4-fluorophenyl methyl-4-fluorobenzene
2723773
1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F
This item is not returnable. View return policy
27064-94-4
C13H9ClF2
238.662
MFCD00044329
FHPNLCLHMNPLEW-UHFFFAOYSA-N
4,4'-difluorobenzhydryl chloride, 4,4'-chloromethylene bis fluorobenzene, 1,1'-chloromethylene bis 4-fluorobenzene, chlorobis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethylchloride, chloro bis-4-fluorophenyl methane, bis 4-fluorophenyl chloromethane, 4,4'-difluorobenzhydrylchloride, 1-chloro 4-fluorophenyl methyl-4-fluorobenzene, 1-chloro-4-fluorophenyl methyl-4-fluorobenzene
2723773
1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.28
Boiling Point 126°C to 128°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.557
Quantity 5 g
UN Number UN1760
Beilstein 2052680
Sensitivity Moisture sensitive
Formula Weight 238.66
Percent Purity 98%
Chemical Name or Material 4,4'-Difluorobenzhydryl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'