missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobenzhydryl chloride, 98%

Product Code. 11463554
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463554 10 g 10g
11399439 5 g 5g
11473554 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463554 Supplier Thermo Scientific Alfa Aesar Supplier No. A11226.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27064-94-4 | C13H9ClF2 | 238.662 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27064-94-4
Molecular Formula C13H9ClF2
Molecular Weight (g/mol) 238.662
MDL Number MFCD00044329
InChI Key FHPNLCLHMNPLEW-UHFFFAOYSA-N
Synonym 4,4'-difluorobenzhydryl chloride, 4,4'-chloromethylene bis fluorobenzene, 1,1'-chloromethylene bis 4-fluorobenzene, chlorobis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethylchloride, chloro bis-4-fluorophenyl methane, bis 4-fluorophenyl chloromethane, 4,4'-difluorobenzhydrylchloride, 1-chloro 4-fluorophenyl methyl-4-fluorobenzene, 1-chloro-4-fluorophenyl methyl-4-fluorobenzene
PubChem CID 2723773
IUPAC Name 1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F

Specifications

Density 1.28
Boiling Point 126°C to 128°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.557
Quantity 10 g
UN Number UN1760
Beilstein 2052680
Sensitivity Moisture sensitive
Formula Weight 238.66
Percent Purity 98%
Chemical Name or Material 4,4'-Difluorobenzhydryl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.