missing translation for 'onlineSavingsMsg'
Learn More

4,4-Diethylamino-2-butyn-1-ol, 98%

Product Code. 15470388
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470388 5 g 5g
15480388 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470388 Supplier Thermo Scientific Alfa Aesar Supplier No. H53501.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10575-25-4 | C8H15NO | 141.21 g/mol

4,4-Diethylamino-2-butyn-1-ol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4-Diethylamino-2-butyn-1-ol is used as pharmaceutical intermediate.

Solubility
Soluble in chloroform and ethyl acetate. Soluble in water 11 g/L @25°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10575-25-4
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.21
MDL Number MFCD00671356
InChI Key ACGZBRWTWOZSFU-UHFFFAOYSA-N
PubChem CID 82735
IUPAC Name 4-(diethylamino)but-2-yn-1-ol
SMILES CCN(CC)CC#CCO

Specifications

Density 0.952
Boiling Point 231°C to 232°C
Flash Point 104°C (219°F)
Linear Formula (CH3CH2)2NCH2C≡CCH2OH
Refractive Index 1.479
Quantity 5 g
Synonym 4-diethylamino but-2-yn-1-ol, 4-diethylamino-2-butyn-1-ol, unii-ma2de62u5w, ma2de62u5w, 2-butyn-1-ol,4-diethylamino, 2-butyn-1-ol, 4-diethylamino, acmc-2098hk, 4-diethylamino-2-butyne-ol, 1-diethylaminobut-2-yn-4-ol
Solubility Information Soluble in chloroform and ethyl acetate. Soluble in water 11g/L @25°C.
Formula Weight 141.21
Percent Purity 98%
Chemical Name or Material 4,4-Diethylamino-2-butyn-1-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.